[1-(2,5-Dimethylbenzyl)piperidin-4-yl]methylamine

Supplier: Matrix Scientific

038457-500MG
101845-544EA 290.77 USD
101845-544
[1-(2,5-Dimethylbenzyl)piperidin-4-yl]methylamine
[1-(2,5-Dimethylbenzyl)piperidin-4-yl]methylamine

Hazard: Irritant

Formula: C₁₅H₂₄N₂
MDL Number: MFCD12027342
CAS Number: 1104927-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR