N-[2-(4-Benzylpiperazin-1-yl)-2-oxoethyl]-N-methylamine

Supplier: Matrix Scientific

038229-500MG
101845-076EA 193.85 USD
101845-076
N-[2-(4-Benzylpiperazin-1-yl)-2-oxoethyl]-N-methylamine
N-[2-(4-Benzylpiperazin-1-yl)-2-oxoethyl]-N-methylamine

Hazard: Irritant

Formula: C₁₄H₂₁N₃O
MDL Number: MFCD06371774
CAS Number: 900641-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR