N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide

Supplier: Matrix Scientific

Synonyms: N-(2,6-Dimethylphenyl)-1-piperazineacetamide

038200-500MG
101845-014EA 290.77 USD
101845-014
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide

Hazard: Irritant

Formula: C₁₄H₂₁N₃O
MW: 247.34 g/mol
MDL Number: MFCD06384973
CAS Number: 5294-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR