2-(4-Methoxyphenyl)-6-methylimidazo[1,2-a]pyridine-3-carbaldehyde

Supplier: Matrix Scientific

038083-500MG
101844-780EA 387.69 USD
101844-780
2-(4-Methoxyphenyl)-6-methylimidazo[1,2-a]pyridine-3-carbaldehyde
2-(4-Methoxyphenyl)-6-methylimidazo[1,2-a]pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₂
MDL Number: MFCD04117111
CAS Number: 727975-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR