3-Amino-3-(4-chlorophenyl)propionic acid

Supplier: Matrix Scientific

Synonyms: 3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(4-chlorophenyl)propionic Acid, 3-Amino-3-(p-chlorophenyl)propionic acid

037904-1G
101844-420EA 23.08 USD
101844-420
3-Amino-3-(4-chlorophenyl)propionic acid
3-Amino-3-(4-chlorophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
MDL Number: MFCD00126370
CAS Number: 19947-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR