3-Amino-1-Boc-piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

037709-5G
101843-998EA 36.92 USD
101843-998
3-Amino-1-Boc-piperidine ≥95%
3-Amino-1-Boc-piperidine

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD01861219
CAS Number: 184637-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR