4'-Methoxybutyrophenone for synthesis

Supplier: Matrix Scientific

Synonyms: 1-(4-Methoxyphenyl)butan-1-one

037431-500MG
101843-360EA 146.15 USD
101843-360
4'-Methoxybutyrophenone for synthesis
4'-Methoxybutyrophenone
Formula: C₁₁H₁₄O₂
Boiling Pt: 120 °C (1 mmHg)
Melting Pt: 26 °C
Density: 1.0451 g/cm³ (20 °C)
MDL Number: MFCD00027138
CAS Number: 4160-51-4
EINECS: 223-995-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR