tert-Butyl-2-piperidin-3-ylethylcarbamate

Supplier: Matrix Scientific

037397-500MG
101843-276EA 278.46 USD
101843-276
tert-Butyl-2-piperidin-3-ylethylcarbamate
tert-Butyl-2-piperidin-3-ylethylcarbamate

Hazard: Irritant

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD01318364
CAS Number: 215305-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR