[4-(Methylsulfonyl)morpholin-2-yl]methylamine hydrochloride

Supplier: Matrix Scientific

037358-500MG
101843-198EA 307.69 USD
101843-198
[4-(Methylsulfonyl)morpholin-2-yl]methylamine hydrochloride
[4-(Methylsulfonyl)morpholin-2-yl]methylamine hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₃S
MDL Number: MFCD12027118
CAS Number: 1185301-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR