N-Cyclohexylpiperidin-4-amine dihydrochloride

Supplier: Matrix Scientific

037311-500MG
101843-104EA 193.85 USD
101843-104
N-Cyclohexylpiperidin-4-amine dihydrochloride
N-Cyclohexylpiperidin-4-amine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD09759078
CAS Number: 1452484-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR