2-[(1-Methylpiperidin-4-yl)oxy]aniline

Supplier: Matrix Scientific

037245-500MG
101842-974EA 387.69 USD
101842-974
2-[(1-Methylpiperidin-4-yl)oxy]aniline
2-[(1-Methylpiperidin-4-yl)oxy]aniline

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD04970986
CAS Number: 869943-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR