Ethyl-1-(cyanomethyl)piperidine-3-carboxylate

Supplier: Matrix Scientific

037046-500MG
101842-564EA 290.77 USD
101842-564
Ethyl-1-(cyanomethyl)piperidine-3-carboxylate
Ethyl-1-(cyanomethyl)piperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂
MDL Number: MFCD09947881
CAS Number: 946702-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR