N-(2-Methoxyethyl)guanidine compound with sulfuric acid (2:1)

Supplier: Matrix Scientific

037018-500MG
101842-512EA 387.69 USD
101842-512
N-(2-Methoxyethyl)guanidine compound with sulfuric acid (2:1)
N-(2-Methoxyethyl)guanidine compound with sulfuric acid

Hazard: Irritant

Formula: C₈H₂₄N₆O₆S
MDL Number: MFCD12027042
CAS Number: 108712-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR