4-Methyl-2-phenyl-5-(pyridin-3-ylmethyl)-1H-pyrazolo[4,3-c]pyridine-3,6(2H,5H)-dione

Supplier: Matrix Scientific

036964-500MG
101842-424EA 335.38 USD
101842-424
4-Methyl-2-phenyl-5-(pyridin-3-ylmethyl)-1H-pyrazolo[4,3-c]pyridine-3,6(2H,5H)-dione
4-Methyl-2-phenyl-5-(pyridin-3-ylmethyl)-1H-pyrazolo[4,3-c]pyridine-3,6(2H,5H)-dione

Hazard: Irritant

Formula: C₁₉H₁₆N₄O₂
MDL Number: MFCD06755313
CAS Number: 847572-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR