5-(Phenylethynyl)-2-furoic acid

Supplier: Matrix Scientific

036878-500MG
101842-260EA 193.85 USD
101842-260
5-(Phenylethynyl)-2-furoic acid
5-(Phenylethynyl)-2-furoic acid

Hazard: Irritant

Formula: C₁₃H₈O₃
MDL Number: MFCD00541949
CAS Number: 130423-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR