[5-(1,3-Benzodioxol-5-yl)-1,2,4-oxadiazol-3-yl]methylamine hydrochloride

Supplier: Matrix Scientific

036857-500MG
101842-218EA 461.54 USD
101842-218
[5-(1,3-Benzodioxol-5-yl)-1,2,4-oxadiazol-3-yl]methylamine hydrochloride
[5-(1,3-Benzodioxol-5-yl)-1,2,4-oxadiazol-3-yl]methylamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₀ClN₃O₃
MDL Number: MFCD12027003
CAS Number: 1119452-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR