1-(Thien-2-ylcarbonyl)piperidin-3-amine hydrochloride

Supplier: Matrix Scientific

036786-500MG
101842-080EA 484.62 USD
101842-080
1-(Thien-2-ylcarbonyl)piperidin-3-amine hydrochloride
1-(Thien-2-ylcarbonyl)piperidin-3-amine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂OS
MW: 246.76 g/mol
MDL Number: MFCD12026977
CAS Number: 1114596-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR