1-(1,3-Benzodioxol-5-ylmethyl)piperidin-4-amine dihydrochloride

Supplier: Matrix Scientific

036670-500MG
101841-846EA 387.69 USD
101841-846
1-(1,3-Benzodioxol-5-ylmethyl)piperidin-4-amine dihydrochloride
1-(1,3-Benzodioxol-5-ylmethyl)piperidin-4-amine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀Cl₂N₂O₂
MDL Number: MFCD12026962
CAS Number: 76167-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR