4-Amino-N-phenylpiperidine-1-carboxamide hydrochloride

Supplier: Matrix Scientific

036640-500MG
101841-786EA 387.69 USD
101841-786
4-Amino-N-phenylpiperidine-1-carboxamide hydrochloride
4-Amino-N-phenylpiperidine-1-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃O
MW: 255.75 g/mol
MDL Number: MFCD08729305
CAS Number: 937602-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR