4-Fluoro-3-[5-(phenoxymethyl)-1,2,4-oxadiazol-3-yl]benzaldehyde

Supplier: Matrix Scientific

036230-500MG
101840-944EA 484.62 USD
101840-944
4-Fluoro-3-[5-(phenoxymethyl)-1,2,4-oxadiazol-3-yl]benzaldehyde
4-Fluoro-3-[5-(phenoxymethyl)-1,2,4-oxadiazol-3-yl]benzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₁FN₂O₃
MDL Number: MFCD12026825
CAS Number: 1119452-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR