N-[(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)methyl]-N-methylamine hydrochloride

Supplier: Matrix Scientific

036180-500MG
101840-842EA 146.15 USD
101840-842
N-[(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)methyl]-N-methylamine hydrochloride
N-[(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)methyl]-N-methylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClN₃O
MDL Number: MFCD12026788
CAS Number: 1185299-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR