1-[5-(Aminosulfonyl)-1,3-benzoxazol-2-yl]piperidine-4-carboxylic acid

Supplier: Matrix Scientific

035879-500MG
101840-250EA 484.62 USD
101840-250
1-[5-(Aminosulfonyl)-1,3-benzoxazol-2-yl]piperidine-4-carboxylic acid
1-[5-(Aminosulfonyl)-1,3-benzoxazol-2-yl]piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₅S
MDL Number: MFCD09701665
CAS Number: 1035841-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR