N-Phthaloylglycine

Supplier: Matrix Scientific

Synonyms: (1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid

035821-100G
101840-136EA 72.31 USD
101840-136
N-Phthaloylglycine
N-Phthaloylglycine

Hazard: Irritant

Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
MDL Number: MFCD00005900
CAS Number: 4702-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR