Littauer Tapered Cutting Forceps, OR Grade, Sklar
Supplier: Sklar
97-1174
10184-734EA 324.26 USD
10184-734
Littauer Tapered Cutting Forceps, OR Grade, Sklar
Bone Cutters and Rongeurs
Sklar's® Littauer Tapered Cutting Forceps are a double spring, plier instrument often used in podiatry as a nail nipper.The tapered blades form a narrow profile and sharp tips useful for accessing difficult to reach sections of a nail.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.