Sklar Blue™ Mini Emmett Hook, OR Grade, Sklar
Supplier: Sklar
91-5246
10184-498EA 405.37 USD
10184-498
Sklar Blue™ Mini Emmett Hook, OR Grade, Sklar
Retractors
This is also be known as an Emmett iris hook, is most commonly used in electrosurgical OB/GYN procedures involving the uterus.
  • The hook is at the end of a long, narrow handle
  • Feature high quality stainless steel with an insulated coating
This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ instrument is fully autoclavable and may also be cleaned ultrasonically.
Sklar Blue™ coated instrument comply with biological testing requirements for cytotoxicity and sensitization regulatory. This also comply with the pre-vacuum and gravity cycle steam sterilization validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coating are guaranteed for one year against manufacturing defects.
Certifications: Meets ISO 10993 requirements.