Sklar Black™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-9108
10184-284EA 1311.98 USD
10184-284
Sklar Black™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Specula
Sklar Black™ Auvard Weighted Vaginal Speculum is a single-end vaginal speculum commonly used in laser OB/GYN procedures
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
It is available with various blade styles and three weight choices in order to accommodate the needs of the patient and the procedure. The weight holds the speculum in place and provides an open view for the surgeon to perform procedures
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects. This product has a 2¾ x 1½" blade with a medium, 2½ pound weight. It has a loop to hold and direct disposable smoke evacuation tubing.