Kevorkian Micro-Cut Biopsy Punch Canula and Tip, OR Grade, Sklar
Supplier: Sklar
90-8976
10184-204EA 1365.39 USD
10184-204
Kevorkian Micro-Cut Biopsy Punch Canula and Tip, OR Grade, Sklar
Biopsy Punches
Sklar's Kevorkian Micro-Cut Biopsy Punch Canula and Tip is part of an OB/GYN instrument
This product attaches to a universal style handle to create biopsy punch forceps. They are used in colposcopy procedures to obtain cervical tissue samples for biopsy. The bite of this Kevorkian punch is 3mm x 8mm and it produces an rectangular shaped tissue sample. It features four lower teeth, useful for grasping and holding difficult to reach surface tissue; however, are not optimal when a deep tissue sample is desired. The length of this product, which is the cannula and the tip only, is approximately 8 inches.