Mini Tischler Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-8964
10184-196EA 1987.55 USD
10184-196
Mini Tischler Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Mini Tischler Micro-Cut Universal Style Biopsy Punch Forceps are an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy
The mini Tischler bite, 2.3mm x 4.2mm, is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred because there is typically less bleeding and pain when the smaller tissue sample is removed. This product is a complete instrument which consists of a handle and a detachable cannula/tip so other universal tip styles may be used with one handle.