Dietary Fibre Extractors, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
F30440210
10184-168EA 14208.18 USD
10184-168
Dietary Fibre Extractors, VELP Scientifica
Extractors Fiber Determination Systems
The CSF 6 filtration unit carries out the final filtration and washing phase required by the enzymatic method for dietary fibre determination.
  • Glass funnels facilitate the transfer of the digested sample and solvents
  • Robust epoxy painted stainless steel housing
  • Short filtration times
Max. number of samples: 6
Ordering information: CSF6 supplied with six glass crucibles with porosity P2