Automatic Kjeldahl Digestion Units, DKL Series
Supplier: VELP SCIENTIFIC INC.
S30100180 S30100190 S30100200 S30100210 S30110190 S30110200
10184-084EA 22132.25 USD
10184-084 10184-086 10184-088 10184-090 10184-092 10184-094
Automatic Kjeldahl Digestion Units, DKL Series
Digestion Systems Mineralization Workstations
Fully automatic Kjeldahl digestion units for nitrogen analysis and protein determination that incorporate the revolutionary TEMS™ technology for unprecedented savings in terms of time, energy - as much as 35%, money and space.
  • Outstanding reliability
  • Automated operation
  • High technology
  • Practical functionality
Outstanding reliability - aluminum block for absolute temperature homogeneity. Unmatched conductivity for fast response up to 450 °C. Conforms to Good laboratory practice standard.
Ordering information:
Combine DKL series with the KS 1000 scrubber for applications where the digestion process produces dangerous fumes (two KS 1000 Scrubbers are recommended for DKL 42/26). Contact your Avantor representative for more information. DKL Series includes the following operating accessories required: lift, suction cap with drip tray, sample rack, test tubes (adequate quantity).