N-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035741-100MG 035741-500MG
101839-892EA 364.62 USD
101839-892 101839-894
N-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)pivalamide
N-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₆H₂₄BClN₂O₃
MW: 338.65 g/mol
MDL Number: MFCD11857740
CAS Number: 1309980-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR