Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
035668-5G
035668-25G
101839-614EA
232.31
USD
101839-614
101839-616
Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥97%
Ethyl 3-amino-1H-pyrazole-4-carboxylate
Hazard: Irritant
Formula:
C₆H₉N₃O₂ Melting Pt: 104…107 °C |
MDL Number:
MFCD00005238 CAS Number: 6994-25-8 |
Specification Test Results
Melting point | 104-107°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...