4-Hydroxyphenylboronic acid ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

035606-5G 035606-25G
101839-560EA 40 USD
101839-560 101839-562
4-Hydroxyphenylboronic acid ≥97% for synthesis
4-Hydroxyphenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₇BO₃
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR