4-Chloro-2-(trifluoromethyl)phenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane

035570-5G
101839-520EA 109.23 USD
101839-520
4-Chloro-2-(trifluoromethyl)phenylboronic acid
4-Chloro-2-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
MDL Number: MFCD04973086
CAS Number: 313545-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR