2-Methoxy-5-(trifluoromethyl)phenylboronic acid
Supplier: Matrix Scientific
Synonyms:
2-Methoxy-5-(trifluoromethyl)benzeneboronic acid
035469-1G
035469-5G
035469-25G
101839-450EA
36.92
USD
101839-450
101839-452
102663-674
2-Methoxy-5-(trifluoromethyl)phenylboronic acid
2-Methoxy-5-(trifluoromethyl)phenylboronic acid
Hazard: Irritant/Store in Freezer
Formula:
C₈H₈BF₃O₃ |
MDL Number:
MFCD04039222 CAS Number: 240139-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...