2,4-Difluoro-3-cyanophenylboronic acid ≥95%

Supplier: Matrix Scientific

035305-1G
101839-360EA 84.62 USD
101839-360
2,4-Difluoro-3-cyanophenylboronic acid ≥95%
2,4-Difluoro-3-cyanophenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₇H₄BF₂NO₂
MDL Number: MFCD12026699
CAS Number: 871940-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR