5-Methoxy-N-phenylnicotinamide

Supplier: Matrix Scientific

035204-250MG
101839-186EA 610.77 USD
101839-186
5-Methoxy-N-phenylnicotinamide
5-Methoxy-N-phenylnicotinamide
Formula: C₁₃H₁₂N₂O₂
MW: 228.25 g/mol
MDL Number: MFCD11857643
CAS Number: 1138443-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR