2-Bromo-N-[3-(2-ethoxyethoxy)phenyl]acetamide

Supplier: Matrix Scientific

035014-500MG
101838-816EA 306.15 USD
101838-816
2-Bromo-N-[3-(2-ethoxyethoxy)phenyl]acetamide
2-Bromo-N-[3-(2-ethoxyethoxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₃
MDL Number: MFCD12026594
CAS Number: 1138442-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR