2-Bromo-N-[2-(1-piperidinylcarbonyl)phenyl]acetamide

Supplier: Matrix Scientific

034963-500MG
101838-714EA 306.15 USD
101838-714
2-Bromo-N-[2-(1-piperidinylcarbonyl)phenyl]acetamide
2-Bromo-N-[2-(1-piperidinylcarbonyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₄H₁₇BrN₂O₂
MDL Number: MFCD12026545
CAS Number: 1138445-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR