2-Bromo-N-(3,5-dichlorophenyl)acetamide

Supplier: Matrix Scientific

034935-500MG
101838-658EA 306.15 USD
101838-658
2-Bromo-N-(3,5-dichlorophenyl)acetamide
2-Bromo-N-(3,5-dichlorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₆BrCl₂NO
MDL Number: MFCD00173816
CAS Number: 57339-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR