N,3,3-Trimethyl-1-butanamine hydrochloride

Supplier: Matrix Scientific

034883-500MG
101838-556EA 338.46 USD
101838-556
N,3,3-Trimethyl-1-butanamine hydrochloride
N,3,3-Trimethyl-1-butanamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₈ClN
MW: 151.68 g/mol
MDL Number: MFCD11226480
CAS Number: 1135288-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR