1-(1,1-Difluoropropyl)-2-fluorobenzene ≥95%

Supplier: Matrix Scientific

034824-1G
101838-442EA 144.62 USD
101838-442
1-(1,1-Difluoropropyl)-2-fluorobenzene ≥95%
1-(1,1-Difluoropropyl)-2-fluorobenzene

Hazard: Irritant

Formula: C₉H₉F₃
MW: 174.17 g/mol
MDL Number: MFCD11845838
CAS Number: 1138445-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR