3,5-Difluoropyridine-2-carbonitrile ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 3,5-Difluoropicolinonitrile, 2-Cyano-3,5-difluoropyridine

034749-1G 034749-5G
101838-282EA 21.54 USD
101838-282 101838-284
3,5-Difluoropyridine-2-carbonitrile ≥97% for synthesis
3,5-Difluoropyridine-2-carbonitrile

Hazard code(s): Irritant-Harmful, Refrigerate

Formula: C₆H₂F₂N₂
Melting Pt: 32…34 °C
MDL Number: MFCD03788759
CAS Number: 298709-29-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 32-34°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR