C-[5-(4-Chlorophenyl)[1,2,4]oxadiazol-3-yl]methylamine hydrochloride

Supplier: Matrix Scientific

033587-500MG
101835-976EA 461.54 USD
101835-976
C-[5-(4-Chlorophenyl)[1,2,4]oxadiazol-3-yl]methylamine hydrochloride
C-[5-(4-Chlorophenyl)[1,2,4]oxadiazol-3-yl]methylamine hydrochloride

Hazard: Irritant

Formula: C₉H₉Cl₂N₃O
MDL Number: MFCD08733174
CAS Number: 1179370-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR