4-(1-Methylpiperidin-4-yl)aniline

Supplier: Matrix Scientific

033049-500MG
101834-914EA 461.54 USD
101834-914
4-(1-Methylpiperidin-4-yl)aniline
4-(1-Methylpiperidin-4-yl)aniline

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MDL Number: MFCD06804550
CAS Number: 454482-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR