1,3,5-Trifluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032564-1G
101834-006EA 256.92 USD
101834-006
1,3,5-Trifluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%
1,3,5-Trifluoro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₄F₆
MDL Number: MFCD11226567
CAS Number: 1099597-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR