3-(Tetrahydro-2-furanylmethoxy)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline
Supplier: Matrix Scientific
032266-500MG
101833-356EA
306.15
USD
101833-356
3-(Tetrahydro-2-furanylmethoxy)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline
3-(Tetrahydro-2-furanylmethoxy)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline
Hazard: Irritant
Formula:
C₂₃H₂₉NO₄ |
MDL Number:
MFCD10688266 CAS Number: 1040682-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...