N-[4-(2-Phenoxyethoxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

032258-500MG
101833-340EA 306.15 USD
101833-340
N-[4-(2-Phenoxyethoxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline
N-[4-(2-Phenoxyethoxy)benzyl]-3-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₂₆H₂₉NO₄
MDL Number: MFCD10688258
CAS Number: 1040681-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR