N-(4-Fluorobenzyl)-3-(tetrahydro-2-furanylmethoxy)aniline
Supplier: Matrix Scientific
032254-500MG
101833-332EA
306.15
USD
101833-332
N-(4-Fluorobenzyl)-3-(tetrahydro-2-furanylmethoxy)aniline
N-(4-Fluorobenzyl)-3-(tetrahydro-2-furanylmethoxy)aniline
Hazard: Irritant
Formula:
C₁₈H₂₀FNO₂ |
MDL Number:
MFCD10688254 CAS Number: 1040681-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...