N-[4-(2,4-Dichlorophenoxy)butyl]-N-(3-methoxypropyl)amine
Supplier: Matrix Scientific
032244-500MG
101833-312EA
306.15
USD
101833-312
N-[4-(2,4-Dichlorophenoxy)butyl]-N-(3-methoxypropyl)amine
N-[4-(2,4-Dichlorophenoxy)butyl]-N-(3-methoxypropyl)amine
Hazard: Irritant
Formula:
C₁₄H₂₁Cl₂NO₂ |
MDL Number:
MFCD10688244 CAS Number: 1040680-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...